Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMC1 Antibody (2H12/4), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB1002617G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002617G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002617G Supplier Novus Biologicals Supplier No. NB1002617G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DMC1 Monoclonal antibody specifically detects DMC1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen DMC1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2H12/4
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Gene Alias disrupted meiotic cDNA1, yeast, homolog of, DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologousrecombination (yeast), DMC1 homologue, HsLim15, LIM15yeast homolog) meiosis-specific homologousrecombination, meiotic recombination protein DMC1/LIM15 homolog, MGC150472, MGC150473
Gene Symbols DMC1
Host Species Mouse
Immunogen Human DMC1 protein (expressed as 6xHis fusion in E. coli and band cut out of gel of insoluble pelet)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Homologous Recombination, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 11144
Test Specificity Specific for DMC1 - does not cross-react with the related protein Rad51.
Target Species Human, Mouse, Bovine, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.