Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DMCM hydrochloride
GREENER_CHOICE

Catalog No. p-4783912
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Benzodiazepine inverse agonist

Benzodiazepine inverse agonist that displays anxiogenic and potent convulsant activity.

Chemical Identifiers

CAS 1215833-62-7
Molecular Formula C17H19ClN2O4

Specifications

Quantity 10 mg
Formula Weight 350.8
Purity >98%
Molecular Weight (g/mol) 350.799
Chemical Name or Material DMCM hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.