Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMP-1 Antibody (001), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB068989 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB068989 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB068989 Supplier Novus Biologicals Supplier No. NBP290020DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DMP-1 Monoclonal specifically detects DMP-1 in Human samples. It is validated for ELISA.

Specifications

Antigen DMP-1
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ARHP, ARHR, dentin matrix acidic phosphoprotein, dentin matrix acidic phosphoprotein 1, Dentin matrix protein 1, DMP-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human DMP-1 (Q13316-1; Met1-Tyr513).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1758
Test Specificity No cross-reactivity in ELISA with: Human CD44 Human CFH
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.