Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMPO Nitrone Adduct Antibody (1), Novus Biologicals™
SDP

Catalog No. NB036586 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036586 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036586 Supplier Novus Biologicals Supplier No. NBP288995
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DMPO Nitrone Adduct Monoclonal specifically detects DMPO Nitrone Adduct in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen DMPO Nitrone Adduct
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 1
Conjugate Unconjugated
Formulation PBS, pH 7.4
Gene Alias 2,2-dimethyl-1-oxido-3,4-dihydropyrrol-1-ium, 2,2-Dimethyl-3,4-dihydro-2H-pyrrole 1-oxide, 5,5-Dimethyl-1-pyrroline N-oxide, 5,5-Dimethyl-1-pyrroline-1-oxide
Host Species Mouse
Immunogen DMPO-octanoic acid conjugated to ovalbumin.
Purification Method Protein G purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.