Learn More
DMPO Nitrone Adduct Antibody (N1664A), DyLight 680, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | DMPO Nitrone Adduct |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | N1664A |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | 2,2-dimethyl-1-oxido-3,4-dihydropyrrol-1-ium, 2,2-Dimethyl-3,4-dihydro-2H-pyrrole 1-oxide, 5,5-Dimethyl-1-pyrroline N-oxide, 5,5-Dimethyl-1-pyrroline-1-oxide |
| Host Species | Mouse |
| Immunogen | 5,5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated to Ovalbumin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.