Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMRT1 Antibody (OTI1G10), Biotin, Novus Biologicals™
SDP

Catalog No. NBP270582B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270582B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270582B Supplier Novus Biologicals Supplier No. NBP270582B

Mouse Monoclonal Antibody

DMRT1 Monoclonal specifically detects DMRT1 in Human samples. It is validated for Western Blot.

Specifications

Antigen DMRT1
Applications Western Blot
Classification Monoclonal
Clone OTI1G10
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias DM domain expressed in testis 1, DMT1DM domain expressed in testis protein 1, doublesex and mab-3 related transcription factor 1, doublesex- and mab-3-related transcription factor 1
Gene Symbols DMRT1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-210 of human DMRT1 (NP_068770) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1761.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.