Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMRT1 Antibody (OTI1G10), PerCP, Novus Biologicals™
SDP

Catalog No. NB289493 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289493 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289493 Supplier Novus Biologicals Supplier No. NBP270582PCP

Mouse Monoclonal Antibody

DMRT1 Monoclonal antibody specifically detects DMRT1 in Human samples. It is validated for Western Blot

Specifications

Antigen DMRT1
Applications Western Blot
Classification Monoclonal
Clone OTI1G10
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias DM domain expressed in testis 1, DMT1DM domain expressed in testis protein 1, doublesex and mab-3 related transcription factor 1, doublesex- and mab-3-related transcription factor 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-210 of human DMRT1(NP_068770) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1761
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.