Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMRTA2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP18001920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18001920 20 μL
NBP180019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18001920 Supplier Novus Biologicals Supplier No. NBP18001920UL

Rabbit Polyclonal Antibody

DMRTA2 Polyclonal specifically detects DMRTA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DMRTA2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. XP_027162
Gene Alias DMRT5, DMRT-like family A2, doublesex- and mab-3-related transcription factor 5, doublesex- and mab-3-related transcription factor A2
Gene Symbols DMRTA2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal of human DMRTA2. Peptide sequence FSGSPLARASKEAGGSCPGSAGPGGGGEEDSPGSASPLGSESGSEADKEE.
Molecular Weight of Antigen 49 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63950
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.