Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ DMSO, sterile filtered



Click to view available options
Quantity:
25 mL
100 mL
Description
DMSO, sterile filtered is a solvent with wide ranging applications in cell culture, cryopreservation and organic chemistry. DMSO dissolves both polar and nonpolar compounds. Improves responsiveness of human embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) to differentiation signals. DMSO is used in PCR to inhibit secondary structure formation by DNA template or primers.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Quantity | 100 mL |
Formula Weight | 78.13 |
Purity | >99.9% |
Molecular Weight (g/mol) | 78.13 |
Chemical Name or Material | DMSO, sterile filtered |
Safety and Handling
Recommended Storage | Store at RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction