Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Antibody (11B6), Novus Biologicals™
SDP

Catalog No. NB1001647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001647 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001647 Supplier Novus Biologicals Supplier No. NB1001647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA Monoclonal specifically detects DNA in All species samples. It is validated for Dot Blot, ELISA, In-situ Hybridization.

Specifications

Antigen DNA
Applications Dot Blot, ELISA, In Situ Hybridization (ISH)
Classification Monoclonal
Clone 11B6
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA
Gene Alias Double stranded deoxyribonucleic acid, Double stranded DNA, dsDNA
Host Species Mouse
Immunogen calf dsDNA
Purification Method Unpurified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Test Specificity In ELISA : This shows 100% reactivity to calf dsDNA and 5% to calf ssDNA. It does not react with RNA.
Reconstitution Reconstitute with 1.0 ml distilled water.
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.