Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Helicase B (HELB) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | DNA Helicase B (HELB) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE2600-25UL
|
MilliporeSigma
ABE260025UL |
25 μL |
Each for $192.35
|
|
|||||
ABE2600-100
|
MilliporeSigma
ABE2600100UL |
100 μL |
Each for $452.71
|
|
|||||
Description
Anti-DNA Helicase B (HELB) Antibody is a highly specific rabbit polyclonal antibody that targets DNA helicase B (HELB) and has been tested for use in Immunocytochemistry and Western Blotting.Specifications
DNA Helicase B (HELB) | |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Epigenetics and Nuclear Function | |
NP_536694 | |
GST-tagged recombinant fragment corresponding to 227 amino acids from the C-terminal half of murine DNA helicase B. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
Helb | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody detects DNA helicase B in murine species. It targets an epitope within 227 amino acids from the C-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title