Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Ligase I Antibody (1A9), HRP, Novus Biologicals™
SDP

Catalog No. NB382211 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382211 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382211 Supplier Novus Biologicals Supplier No. NBP308267H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA Ligase I Monoclonal antibody specifically detects DNA Ligase I in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen DNA Ligase I
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 1A9
Conjugate HRP
Formulation PBS
Gene Alias DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1
Host Species Mouse
Immunogen Full-length native bovine DNA Ligase I protein (Uniprot: P18858)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3978
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.