Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Ligase I Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB048266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB048266 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB048266 Supplier Novus Biologicals Supplier No. NBP297831G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA Ligase I Polyclonal specifically detects DNA Ligase I in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DNA Ligase I
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DNA Ligase I fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3978
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.