Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase I Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056635
Description
DNA Ligase I Polyclonal specifically detects DNA Ligase I in Human samples. It is validated for Western Blot.Specifications
DNA Ligase I | |
Polyclonal | |
Western Blot 1:500-1:1000 | |
P18858 | |
LIG1 | |
This antibody was developed against a synthetic peptide containing amino acids 51-68 of human LIG1. | |
0.2 mL | |
Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
3978 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS containing 0.1 mg/ml BSA. with 0.02% Sodium Azide | |
DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction