Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase I Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | DNA Ligase I |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056635T
![]() |
Novus Biologicals
NB10056635SS |
0.05 mL |
Each for $181.90
|
|
|||||
NB10056635
![]() |
Novus Biologicals
NB10056635 |
0.2 mL |
Each for $402.00
|
|
|||||
Description
DNA Ligase I Polyclonal specifically detects DNA Ligase I in Human samples. It is validated for Western Blot.Specifications
DNA Ligase I | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
PBS containing 0.1 mg/ml BSA. with 0.02% Sodium Azide | |
DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 | |
LIG1 | |
IgG | |
Protein G purified |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
P18858 | |
3978 | |
This antibody was developed against a synthetic peptide containing amino acids 51-68 of human LIG1. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title