Learn More
DNA Ligase I Mouse anti-Human, Mouse, Rat, Bovine, Clone: 1A9, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | DNA Ligase I |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 1A9 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, ELISA, Immunoprecipitation |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.