Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Ligase III Antibody (HL2280) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295176 Supplier Novus Biologicals Supplier No. NBP325441
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DNA Ligase III Monoclonal antibody specifically detects DNA Ligase III in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen DNA Ligase III
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL2280
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DNA ligase 3, DNA ligase III, EC 6.5.1.1, LIG2, ligase II, DNA, ATP-dependent, ligase III, DNA, ATP-dependent, Polydeoxyribonucleotide synthase [ATP] 3
Host Species Rabbit
Immunogen Recombinant fragment of human DNA Ligase III.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 3980
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.