Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Ligase III Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB134632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB134632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB134632 Supplier Novus Biologicals Supplier No. NBP306266H

Rabbit Polyclonal Antibody

DNA Ligase III Polyclonal specifically detects DNA Ligase III in Human samples. It is validated for Western Blot.

Specifications

Antigen DNA Ligase III
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias DNA ligase 3, DNA ligase III, EC 6.5.1.1, LIG2, ligase II, DNA, ATP-dependent, ligase III, DNA, ATP-dependent, Polydeoxyribonucleotide synthase [ATP] 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DNA Ligase III fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 3980
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.