Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Methyltransferase Inhibitor, PROMO, Thermo Scientific Chemicals

Catalog No. J64630MAPM
Click to view available options
Quantity:
10 mg

Chemical Identifiers

CAS 48208-26-0
Molecular Formula C19H14N2O4
Synonym RG108; N-Phthalyl-L-tryptophan

Specifications

Quantity 10 mg
For Use With (Application) A cell-permeable, specific DNA methyltransferases inhibitor
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.