Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA-PKcs Antibody (3H6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222128 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222128 0.1 mL
NBP222128SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222128 Supplier Novus Biologicals Supplier No. NBP222128
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA-PKcs Monoclonal antibody specifically detects DNA-PKcs in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen DNA-PKcs
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3H6
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P78527
Gene Alias DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p460
Gene Symbols PRKDC
Host Species Mouse
Immunogen Purified recombinant fragment of human DNA PKcs expressed in E. coli. [Uniprot: P78527]
Molecular Weight of Antigen 460 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, DNA replication Transcription Translation and Splicing, Non homologous end joining, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5591
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.