Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA-PKcs Antibody (3H6), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337570 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337570 Supplier Novus Biologicals Supplier No. NBP222128MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA-PKcs Monoclonal antibody specifically detects DNA-PKcs in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen DNA-PKcs
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3H6
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p460
Host Species Mouse
Immunogen Purified recombinant fragment of human DNA PKcs expressed in E. coli. [Uniprot: P78527]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, DNA replication Transcription Translation and Splicing, Non homologous end joining, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5591
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.