Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase delta, catalytic subunit Antibody (607), Novus Biologicals™
SDP

Catalog No. NB100476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100476 Supplier Novus Biologicals Supplier No. NB100476
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA Polymerase delta, catalytic subunit Monoclonal specifically detects DNA Polymerase delta, catalytic subunit in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen DNA Polymerase delta, catalytic subunit
Applications Western Blot
Classification Monoclonal
Clone 607
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Gene Alias CDC2, CDC2 homolog, DNA polymerase delta catalytic subunit, DNA polymerase subunit delta p125, DNA-directed polymerase delta 1, EC 2.7.7.7, POLD, polymerase (DNA directed), delta 1, catalytic subunit (125kD), polymerase (DNA directed), delta 1, catalytic subunit 125kDa
Gene Symbols POLD1
Host Species Mouse
Immunogen Recombinant protein encoding amino acids 1-140 of the large subunit of human DNA polymerase d.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5424
Test Specificity DNA Polymerase delta (607). Cross-reacts with Human and Mouse. Not yet tested in other species.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.