Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase epsilon catalytic subunit A Antibody (3C5.1), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB355160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355160 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355160 Supplier Novus Biologicals Supplier No. NB100115DL594

Mouse Monoclonal Antibody

DNA Polymerase epsilon catalytic subunit A Monoclonal antibody specifically detects DNA Polymerase epsilon catalytic subunit A in Human, Mouse, Mustelid, Primate, Drosophila (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen DNA Polymerase epsilon catalytic subunit A
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3C5.1
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp434F222, DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A, EC 2.7.7, EC 2.7.7.7, POLE1FLJ21434, polymerase (DNA directed), epsilon
Host Species Mouse
Immunogen Human DNA Polymerase epsilon purified from HeLa cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Polymerases, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5426
Target Species Human, Mouse, Mustelid, Primate, Drosophila (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.