Learn More
DNA Polymerase epsilon catalytic subunit A Antibody, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | DNA Polymerase epsilon catalytic subunit A |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
| Formulation | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Gene Alias | DKFZp434F222, DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A, EC 2.7.7, EC 2.7.7.7, POLE1FLJ21434, polymerase (DNA directed), epsilon |
| Gene Symbols | POLE |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QGYLITNREIVSEDIEDFEFTPKPEYEGPFCVFNEPDEAHLIQRWFEHVQETKPTIMVTYNGDFFDWPFVEARAAVHGLSMQQEI |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.