Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase epsilon subunit 3 Antibody (PCRP-POLE3-2F10), HRP, Novus Biologicals™
SDP

Catalog No. NB379776 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379776 Supplier Novus Biologicals Supplier No. NBP308571H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA Polymerase epsilon subunit 3 Monoclonal antibody specifically detects DNA Polymerase epsilon subunit 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen DNA Polymerase epsilon subunit 3
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone PCRP-POLE3-2F10
Conjugate HRP
Formulation PBS
Gene Alias Arsenic-transactivated protein, asTP, CHARAC17arsenic transactivated protein, CHRAC-17, CHRAC17Ybl1, Chromatin accessibility complex 17 kDa protein, DNA polymerase epsilon p17 subunit, DNA polymerase epsilon subunit 3, DNA polymerase epsilon subunit p17, DNA polymerase II subunit 3, EC 2.7.7.7, histone fold protein CHRAC17, HuCHRAC17, p17, polymerase (DNA directed), epsilon 3 (p17 subunit), YBL1
Host Species Mouse
Immunogen Recombinant full-length human DNA Polymerase epsilon subunit 3 protein (Uniprot: Q9NRF9)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Polymerases, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 54107
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.