Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase gamma Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB136236 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB136236 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB136236 Supplier Novus Biologicals Supplier No. NBP306345AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA Polymerase gamma Polyclonal specifically detects DNA Polymerase gamma in Human samples. It is validated for Western Blot.

Specifications

Antigen DNA Polymerase gamma
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DNA Polymerase gamma fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Polymerases, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5428
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.