Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase gamma Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095653 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095653 20 μL
NB095652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095653 Supplier Novus Biologicals Supplier No. NBP30382620UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA Polymerase gamma Polyclonal specifically detects DNA Polymerase gamma in Human samples. It is validated for Western Blot.

Specifications

Antigen DNA Polymerase gamma
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 980-1239 of human DNA Polymerase gamma (NP_002684.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline DNA Polymerases, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5428
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.