Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase lambda Antibody, Novus Biologicals™
SDP

Catalog No. NB10081665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10081665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10081665 Supplier Novus Biologicals Supplier No. NB10081665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA Polymerase lambda Polyclonal specifically detects DNA Polymerase lambda in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Western Blot (Negative).

Specifications

Antigen DNA Polymerase lambda
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Western Blot (Negative)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500-1:2000, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500-1:2000, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_037406.1
Gene Alias BETAN, DNA polymerase beta-2, DNA polymerase beta-N, DNA polymerase kappa, DNA polymerase lambda, EC 2.7.7.7, EC 4.2.99.-, FLJ46002, Pol beta2, Pol Lambda, POLKAPPA, polymerase (DNA directed), lambda
Gene Symbols POLL
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human polymerase (DNA directed), lambda using the numbering given in entry NP_037406.1 (GeneID 27343).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 27343
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Whitetufted-ear marmoset, Common squirrel monkey, Crab-eating macaque, Olive baboon and Bolivian red howler monkey.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.