Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase lambda Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | DNA Polymerase lambda |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
DNA Polymerase lambda Polyclonal antibody specifically detects DNA Polymerase lambda in Human samples. It is validated for ImmunofluorescenceSpecifications
DNA Polymerase lambda | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, DNA Polymerases, DNA Repair, Immunology | |
PBS, pH 7.2, 40% glycerol | |
27343 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
BETAN, DNA polymerase beta-2, DNA polymerase beta-N, DNA polymerase kappa, DNA polymerase lambda, EC 2.7.7.7, EC 4.2.99.-, FLJ46002, Pol beta2, Pol Lambda, POLKAPPA, polymerase (DNA directed), lambda | |
This antibody was developed against Recombinant Protein corresponding to amino acids: VVPYSEFACALLYFTGSAHFNRSMRALAKTKGMSLSEHALSTAVVRNTHGCKVGPGRVLPTPTEKDVFRLLGLPYREPAERD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title