Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA polymerase mu Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158202
Description
DNA polymerase mu Polyclonal specifically detects DNA polymerase mu in Human samples. It is validated for Western Blot.Specifications
DNA polymerase mu | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DNA polymerase mu, DNA-directed DNA/RNA polymerase mu, EC 2.7.7.7, FLJ35482, Pol iota, Pol Mu, polmu, polymerase (DNA directed), mu, polymerase (DNA-directed), mu, Tdt-N, Terminal transferase | |
Rabbit | |
55 kDa | |
100 μL | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
27434 | |
Reconstitute in 50μL distilled water to a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9NP87 | |
POLM | |
Synthetic peptides corresponding to POLM(polymerase (DNA directed), mu) The peptide sequence was selected from the middle region of POLM (NP_037416). Peptide sequence LRRFSRKEKGLWLNSHGLFDPEQKTFFQAASEEDIFRHLGLEYLPPEQRN. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Pig: 100%; Guinea pig: 92%; Mouse: 85%; Rabbit: 85%; Rat: 85%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction