Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA polymerase sigma Antibody, Novus Biologicals™
SDP

Catalog No. NB10056356T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056356T 0.025 mL
NB10056356 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056356T Supplier Novus Biologicals Supplier No. NB10056356SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA polymerase sigma Polyclonal specifically detects DNA polymerase sigma in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen DNA polymerase sigma
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500
Formulation PBS with 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_008930.1
Gene Alias DNA polymerase kappa, DNA polymerase sigma, EC 2.7.7.7, LAK-1TUTase 5, PAP associated domain containing 7, POLKTRF41, POLSTUTASE5, polymerase (DNA directed) sigma, polymerase (DNA-directed) sigma, Terminal uridylyltransferase 5, Topoisomerase-related function protein 4-1, TRF4-1LAK1, TRF4PAP-associated domain-containing protein 7
Gene Symbols TENT4A
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide containing amino acids 447-462 (GTASLKAVHHMSSPAI) of human TRF4-1, GenBank no NP_008930.1.
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11044
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.