Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Primase large subunit Antibody, Novus Biologicals™
SDP

Catalog No. NB393424 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393424 25 μL
NBP258498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393424 Supplier Novus Biologicals Supplier No. NBP25849825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA Primase large subunit Polyclonal specifically detects DNA Primase large subunit in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DNA Primase large subunit
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide
Gene Alias dJ422B11.1.1, DNA primase 58 kDa subunit, DNA primase subunit p58, EC 2.7.7, EC 2.7.7.-, MGC75142, p58, polypeptide 2A, p58, PRIM2ADNA primase large subunit, primase polypeptide 2A, 58kDa, primase, DNA, polypeptide 2 (58kDa), primase, polypeptide 2A (58kD), primase, polypeptide 2A, 58kDa
Gene Symbols PRIM2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ASYPHCLQFYLQPPSENISLIEFENLAIDRVKLLKSVENLGVSYVKGTEQYQSKLESELRKLKFSYRENLEDEYEPRRR
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5558
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.