Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA/RNA Damage Antibody (15A3), HRP, Novus Biologicals™
SDP

Catalog No. NB11096878H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096878H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11096878H Supplier Novus Biologicals Supplier No. NB11096878H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA/RNA Damage Monoclonal specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay, Dot Blot.

Specifications

Antigen DNA/RNA Damage
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay, Dot Blot
Classification Monoclonal
Clone 15A3
Conjugate HRP
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay, Dot Blot
Formulation PBS with No Preservative
Host Species Mouse
Immunogen 8-hydroxy-guanosine-BSA and-casein conjugates
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine).
Target Species Human, Mouse, Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.