Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA/RNA Damage Antibody (15A3), Novus Biologicals™
SDP

Catalog No. NB11096878K Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096878K 0.025 mg
NB11096878 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11096878K Supplier Novus Biologicals Supplier No. NB11096878SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 12 publications

DNA/RNA Damage Monoclonal specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoassay, Dot Blot, Functional Assay.

Specifications

Antigen DNA/RNA Damage
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Dot Blot, Functional Assay
Classification Monoclonal
Clone 15A3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay, Dot Blot 1:100-1:200, Functional Assay
Formulation PBS and 50% Glycerol with 0.09% Sodium Azide
Host Species Mouse
Immunogen 8-hydroxy-guanosine-BSA and-casein conjugates
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine).
Target Species Human, Mouse, Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.