Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA/RNA Damage Antibody (15A3), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 12 publications
$243.50 - $584.50
Specifications
Antigen | DNA/RNA Damage |
---|---|
Clone | 15A3 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay, Dot Blot 1:100-1:200, Functional Assay |
Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Dot Blot, Functional Assay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11096878K
![]() |
Novus Biologicals
NB11096878SS |
0.025 mg |
Each for $243.50
|
|
|||||
NB11096878
![]() |
Novus Biologicals
NB11096878 |
0.1 mg |
Each for $584.50
|
|
|||||
Description
DNA/RNA Damage Monoclonal specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoassay, Dot Blot, Functional Assay.Specifications
DNA/RNA Damage | |
1 mg/mL | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Dot Blot, Functional Assay | |
Unconjugated | |
Mouse | |
Human, Mouse, Non-species specific | |
IgG2b | |
Protein G purified | |
Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine). |
15A3 | |
Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay, Dot Blot 1:100-1:200, Functional Assay | |
Monoclonal | |
Purified | |
RUO | |
8-hydroxy-guanosine-BSA and-casein conjugates | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title