Learn More
G-Biosciences DNA Size Selection and PCR Clean Up Magnetic Beads
SCI_ED
G-Biosciences DNA Size Selection and PCR Cleanup Magnetic Beads are suitable for PCR clean-up and DNA fragment size selection from 150 bp to 1000 bp and even larger.
Supplier:  G-Biosciences 7862166
 View more versions of this product
Catalog No. S04218
Only null left
Add to Cart
Description
Description
DNA size selection and PCR clean up Magnetic Beads remove excess nucleotides, salts, enzymes, and other impurities with a simple washing process to obtain purified fragments, which can be directly used in downstream applications such as sequencing, hybridization, and enzyme digestion. DNA size selection depends on optimizing magnetic beads, target DNA ratio, and salt concentrations.
Specifications
Specifications
Magnetic Beads
Random
450 mL
Ambion™, Applied Biosystems™, DNAZap™, RNaseAlert™, RNase Away™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.