Learn More
G-Biosciences DNA Size Selection and PCR Clean Up Magnetic Beads
G-Biosciences DNA Size Selection and PCR Cleanup Magnetic Beads are suitable for PCR clean-up and DNA fragment size selection from 150 bp to 1000 bp and even larger.
$256.00 - $5689.00
Specifications
Product Type | Magnetic Beads |
---|---|
Primer | Random |
Product Line | Ambion™, Applied Biosystems™, DNAZap™, RNaseAlert™, RNase Away™ |
Description
DNA size selection and PCR clean up Magnetic Beads remove excess nucleotides, salts, enzymes, and other impurities with a simple washing process to obtain purified fragments, which can be directly used in downstream applications such as sequencing, hybridization, and enzyme digestion. DNA size selection depends on optimizing magnetic beads, target DNA ratio, and salt concentrations.
Specifications
Magnetic Beads | |
Random | |
Ambion™, Applied Biosystems™, DNAZap™, RNaseAlert™, RNase Away™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.