Learn More
Invitrogen™ DNAH8 Polyclonal Antibody

Description
Immunogen sequence: ILNHKSKHVE EAVRELISIF EQIYEVKYTG KVGKQSEQRK HVVFGSETEE GENNDYEANI VNEFDTHDKE DEFKKECKEV FAFFSHQLLD SLQKATRLSL D Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 75%.
Specifications
Specifications
| Antigen | DNAH8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DNAH8 |
| Gene Accession No. | Q96JB1 |
| Gene Alias | ATPase; Axonemal beta dynein heavy chain 8; ciliary dynein heavy chain 8; DNAH8; dynein axonemal heavy chain 8; dynein heavy chain 8, axonemal; dynein, axonemal, heavy chain 8; dynein, axonemal, heavy polypeptide 8; hdhc9 |
| Gene Symbols | DNAH8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.