Learn More
Description
Specifications
Specifications
| Antigen | DNAJA2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI3A10 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | cell cycle progression 3 protein, Cell cycle progression restoration gene 3 protein, CPR3PRO3015, Dj3, DJA2, DNAJ, DnaJ (Hsp40) homolog, subfamily A, member 2, dnaJ homolog subfamily A member 2, DNJ3, HIRA interacting protein 4, HIRA-interacting protein 4, HIRIP4RDJ2, Renal carcinoma antigen NY-REN-14 |
| Gene Symbols | DNAJA2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
