Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJA4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017294 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017294 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017294 Supplier Novus Biologicals Supplier No. NBP281983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJA4 Polyclonal specifically detects DNAJA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen DNAJA4
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, ELISA
Formulation PBS
Gene Alias DnaJ (Hsp40) homolog, subfamily A, member 4, dnaJ homolog subfamily A member 4, MST104, MSTP104, PRO1472
Gene Symbols DNAJA4
Host Species Rabbit
Immunogen DNAJA4 antibody was raised against a 15 amino acid peptide near the center of human DNAJA4. The immunogen is located within amino acids 210 - 260 of DNAJA4. Amino Acid Squence: HGEGDQEPELEPGDV
Molecular Weight of Antigen 40 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55466
Test Specificity DNAJA4 antibody is human, mouse, and rat reactive. At least three isoforms of DNAJA4 are known to exist; this antibody will detect all three isoforms.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.