Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC13 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341868 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341868 Supplier Novus Biologicals Supplier No. NBP281919PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJC13 Polyclonal antibody specifically detects DNAJC13 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen DNAJC13
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 13, DnaJ domain-containing protein RME-8, dnaJ homolog subfamily C member 13, required for receptor-mediated endocytosis 8
Host Species Rabbit
Immunogen DNAJC13 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human DNAJC13. The immunogen is located within amino acids 1940 - 1990 of DNAJC13. Amino Acid Squence: EHFKNQQDNPEANWKLPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23317
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.