Learn More
Invitrogen™ DNAJC13 Polyclonal Antibody

Description
A recommended positive control is human heart tissue lysate. At least two isoforms of DNAJC13 are known to exist; this antibody will detect both isoforms. DNAJC13 antibody is not expected to cross-react with other DNAJC family members.
Specifications
Specifications
| Antigen | DNAJC13 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | DNAJC13 |
| Gene Accession No. | O75165 |
| Gene Alias | D030002L11Rik; DnaJ (Hsp40) homolog, subfamily C, member 13; DnaJ domain-containing protein RME-8; DnaJ heat shock protein family (Hsp40) member C13; dnaJ homolog subfamily C member 13; DNAJC13; Gm1124; j-domain containing protein, required for endocytosis; KIAA0678; mKIAA0678; PARK21; receptor mediated endocytosis RME-8; required for receptor-mediated endocytosis 8; Rme8; RME-8 |
| Gene Symbols | DNAJC13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.