Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290498 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290498 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290498 Supplier Novus Biologicals Supplier No. NBP212802APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJC2 Polyclonal antibody specifically detects DNAJC2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen DNAJC2
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 2, dnaJ homolog subfamily C member 2, MPHOSPH11M-phase phosphoprotein 11, MPP11Zuotin-related factor 1, ZRF1zuotin, ZUO1, zuotin related factor 1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 500 and 550 of human DnaJ (Hsp40) Homolog, Subfamily C, Member 2 using the numbering given in entry NP_055192.1 (GeneID 27000).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27000
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.