Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJC22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17932820UL
Description
DNAJC22 Polyclonal specifically detects DNAJC22 in Mouse samples. It is validated for Western Blot.Specifications
| DNAJC22 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| NP_789805 | |
| DNAJC22 | |
| Synthetic peptide directed towards the middle region of human 2810451A06Rik. Peptide sequence LVAAVGNQTSDFKNTLGAAFLTSPVFYGRPIAILPISLAASITAQKHRRY. | |
| 20 μL | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| DnaJ (Hsp40) homolog, subfamily C, member 22, dnaJ homolog subfamily C member 22, FLJ13236, wurst homolog, wus | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79962 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction