Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC30 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB064793 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064793 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064793 Supplier Novus Biologicals Supplier No. NBP300191C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJC30 Polyclonal specifically detects DNAJC30 in Human samples. It is validated for ELISA.

Specifications

Antigen DNAJC30
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 30, dnaJ homolog subfamily C member 30, MGC12943, WBSCR18, Williams Beuren syndrome chromosome region 18, Williams-Beuren syndrome chromosomal region 18 protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant DNAJC30 (Q96LL9; Ser39-Gly124).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84277
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.