Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC30 Antibody, Novus Biologicals™
SDP

Catalog No. NB407486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB407486 25ul
NBP180621 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB407486 Supplier Novus Biologicals Supplier No. NBP18062125UL

Rabbit Polyclonal Antibody has been used in 1 publication

DNAJC30 Polyclonal specifically detects DNAJC30 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DNAJC30
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 30, dnaJ homolog subfamily C member 30, MGC12943, WBSCR18, Williams Beuren syndrome chromosome region 18, Williams-Beuren syndrome chromosomal region 18 protein
Gene Symbols DNAJC30
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:YYRQCFLYHPDRNSGSAEAAERFTRISQAYVVLGSATLRRKYDRGLLSDEDLRGPGVRPSRTPAPDPGSPRTPPPTSRTHDGSRASPGANRTMFNFDAFYQAHYGEQLERER
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84277
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.