Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAJC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | DNAJC4 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119173
|
Invitrogen™
PA5119173 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122031
|
Invitrogen™
PA5122031 |
20 μL |
Each for $209.50
|
|
|||||
Description
DNAJC4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
DNAJC4 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Pig | |
DNAJC4 | |
2010301J22Rik; DANJC4; DnaJ (Hsp40) homolog, subfamily C, member 4; DnaJ heat shock protein family (Hsp40) member C4; dnaJ homolog subfamily C member 4; DNAJC4; DnaJ-like protein HSPF2; heat shock 40kD protein 2; Hspf2; MCG18; multiple endocrine neoplasia type 1 candidate protein 18; multiple endocrine neoplasia type 1 candidate protein number 18; Multiple endocrine neoplasia type 1 candidate protein number 18 homolog | |
DNAJC4 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q9NNZ3 | |
100522005, 3338 | |
Recombinant protein Heat Shock 40kDa Protein 2. The antigen corresponds to amino acid range 1-150 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title