Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC5G Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$382.00 - $610.00

Specifications

Antigen DNAJC5G
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP238484
SDP
View Documents
Novus Biologicals
NBP238484
0.1 mL
Each for $610.00
Only null left
Add to Cart
 
NB396806
SDP
View Documents
Novus Biologicals
NBP23848425UL
25 μL
Each for $382.00
Only null left
Add to Cart
 
Description

Description

DNAJC5G Polyclonal specifically detects DNAJC5G in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DNAJC5G
Polyclonal
Rabbit
Human
Q8N7S2
285126
This antibody was developed against a recombinant protein corresponding to amino acids: NAAHAILSDSKKRKIYDQHGSLGIYLYDHFGEEGVRY
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
CSP-gamma, DnaJ (Hsp40) homolog, subfamily C, member 5 gamma, dnaJ homolog subfamily C member 5G, FLJ40417, gamma cysteine string protein, gamma-CSP, Gamma-cysteine string protein
DNAJC5G
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.