Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC8 Antibody, Novus Biologicals™
SDP

Catalog No. NB10097847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10097847 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10097847 Supplier Novus Biologicals Supplier No. NB10097847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJC8 Polyclonal specifically detects DNAJC8 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen DNAJC8
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_055095.2
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 8, dnaJ homolog subfamily C member 8, HSPC331, SPF31Splicing protein spf31
Gene Symbols DNAJC8
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 203 and 253 of human DnaJ (Hsp40) homolog, subfamily C, member 8 using the numbering given in entry NP_055095.2 (GeneID 22826).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22826
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.