Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAM-1/CD226 Antibody (26), Janelia Fluor™ 549
SDP

Catalog No. NB074212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074212 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074212 Supplier Novus Biologicals Supplier No. NBP290489JF549

Rabbit Monoclonal Antibody

DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Mouse samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen DNAM-1/CD226
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 26
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, ELISA
Formulation 50 mM Sodium Borate
Gene Alias CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DNAM-1/CD226 (NP_848802.2; Met 1-Pro 254).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10666
Test Specificity No cross-reactivity in ELISA with: Human CD226/ DNAM-1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.